Learn More
BINDER™ Series ED Classic.Line - Drying and Heating Chambers, with Natural Convection
GREENER_CHOICE
Product Code. 15602086 Shop All BINDER Products
Click to view available options
Capacity (Metric):
20 L
400 L
Uniformity:
2.5°C
3°C
Unit Size:
Each
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
Product Code. 15602086
Brand: BINDER™ 90100075
Only null left
Add to basket
Edge
Add to basket
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
The strengths of a BINDER ED series drying chamber include routine drying and sterilization tasks up to +300 °C. Thanks to the natural convection, all thermal processes which use this drying chamber are highly efficient. The ED series ensures fast, uniform drying.
  • Uniform drying conditions thanks to APT.line™ technology
  • Identical test conditions throughout the chamber interior independent of sample size and quantity
  • Outstanding thermal insulation saves operation costs
  • Temperature range: room temperature plus 5 to 300°C
  • APT.line™ preheating chamber technology
  • Natural convection
  • Adjustable exhaust air flap
  • Controller with timer functions
  • 2 chrome-plated racks
  • Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
Specifications
Certifications/Compliance ASTM D5374, DIN 12880
Description ED400-400 V-I, with RS 422, Series ED Classic.Line - Drying and heating chambers with natural convection, 400 L, 400 V 3 ∼ 50/60 Hz
Capacity (Metric) 400 L
Temperature (Metric) 5°C to 300°C
Type Drying and Heating Chamber
Display LED
Model ED400-400 V-I
No. of Shelves 10 (Maximum), 2 (Standard)
Ports 10/30/50/100 mm
Safety Features User friendly and easy to operate all-digital PID controller technology
Temperature Uniformity 150°C
Timer 0 to 99 hr
Uniformity 3°C
Dimensions (D x W x H) Interior 1000 x 520 x 800 mm
Dimensions (L x W x H) 1290 x 860 x 1050 mm
Width (Metric) 1235 mm
Height (Metric) 1025 mm
Weight (Metric) 125 kg
Voltage 400 V
Wattage 3.4 kW
Amperage 16 A
Frequency 50/60 Hz