Learn More
LABPLAS Dry Sani-Sponge
Biocide free dry sponge for sampling surfaces.
Brand: LABPLAS KSS-82100
This item is not returnable.
View return policy
Description
- SANI-SPONGE
- Dry sponge
- Dimensions: 178 x 305 mm
- Holds: 1650 mL
Specifications
Hygiene Sponge Sampling Kit | |
Sterile |