Learn More
asecos™ Drum Rack With Sump Pallet PE
The Drum Rack With Sump Pallet PE provides safe storage and filling storage of substances hazardous to water of all hazard classes.
Brand:  asecos™ 13147
 View more versions of this product
Product Code. 17816516
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • For the storage of acids, alkalis and further aggressiv chemicals
  • Safe dispensing due to overhanging collecting sump
Specifications
Specifications
15.9 gal.
Steel
29.72 in.
29.72 in.
61.81 in.
31.30 in.
108.03 lb.
60 L
Drum Rack
755.00 mm
755.00 mm
1570.00 mm
795.00 mm
49 kg