Learn More
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Promotion
Product Code. 15707526 Shop All Thermo Scientific Products
Click to view available options
Quantity:
4 x 1.25 mL
20 x 1.25mL
No. of Reactions:
200
1000
Unit Size:
1000 reactions
200 reactions
Product Code. 15707526
Brand: Thermo Scientific™ K1081
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Contains green buffer that includes density reagent and two tracking dyes for direct loading of PCR products on gels
  • DreamTaq DNA Pol,
  • dNTPs,
  • Green buffer, density reagent and dyes for direct loading,
  • supplied with Water,
  • Nuclease free
TRUSTED_SUSTAINABILITY
Specifications
Concentration 2X
Quantity 4 x 1.25 mL
For Use With (Application) Standard PCR, RT-PCR, Genotyping, Cloning
No. of Reactions 200