Learn More
Fisherbrand™ PTFE Magnetic Stir Bar, Double-sided crosshead
Designed for use with tube-like containers yet very effective as general stirrers. The PTFE-coated magnet stirrers include a crosshead on both sides of the stirrer for greater turbulence.
Brand:  Fisherbrand™ 001.1135
 View more versions of this product
Product Code. 16245731
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Stable, general-purpose magnetic stirrer
  • Designed for use with tube-like containers yet very effective as general stirrers
  • Double-sided crossheads provider greater turbulence during mixing
  • Autoclavable
Specifications
Specifications
Autoclavable
White
Containers & Vessels / Stirrer Hot Plates
35 mm
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
12mm
2/Pk.