missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Double Platform Test Tube Rack

Test Tube racks are an effective way of storing Falcon/ Universal tubes, Bijous, cryogenic vials or any size glass bottles.

Brand:  Wesbart™ DP18100

 View more versions of this product

Product Code. 11711714

  • 65.25€ / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Racks are available in aluminum and stainless steel with the option of a hand grip
  • Test Tube racks can be designed to fit in horizontal cold store or be free standing
  • Different size test tubes racks are available to suit your requirements
  • Labels can be fitted to racks so they can be easily identified
  • Specifications

    Specifications

    Double Platform Test Tube Rack
    Autoclavable
    Tubes (18 mm)
    100
    Upright Freezers
    18 mm
    10 x 10
    Natural
    Aluminum
    Non-sterile
    233 x 233 x 60 mm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Promotions

    Promotions