Learn More
Fisherbrand™ Double-Ended PTFE Stir Bar
Provides stable stirring action. These PTFE magnetic stir bars have double paddle action to give efficient stirring plus high stability.
Brand: Fisherbrand™
Additional Details : Weight : 0.08500kg
Description
- Stable, general-purpose magnetic stir bar
- Double paddle action gives efficient stirring plus high stability
- Available in plain or color-coded
- Autoclavable
Specifications
8 mm | |
White / Plain | |
Containers & Vessels / Stirrer Hot Plates | |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
PTFE encapsulated Alnico V magnet core | |
1/Ea. | |
Yes | |
55 mm |