missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Double Bagged Bottled Tryptone Soya Broth (TSB)

Product Code. p-7078332
Click to view available options
Format:
Dbl Bagged Borosilicate Bottle
Double Bagged Serum Bottle
Cap Type:
PP plastic cap w/ Teflon
Stopper, Crimp seal
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17729448

Brand: Thermo Scientific™ R112751

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Perform confident sterility testing with double bagged bottled media for added security between environmental class changes.

Perform confident sterility and microbial limits testing with Thermo Scientific™ Remel™ Double Bagged Bottled Tryptone Soya Broth (TSB) for testing according to USP, JP, and EU regulations and as a general purpose enrichment broth for cultivation and isolation of microorganisms. Double Bagged Bottled Tryptone Soya Broth (TSB) is ideal for microbiological testing that requires additional packaging for sterility protection between environmental class changes.

Add Remel Double Bagged Bottled Tryptone Soya Broth (TSB) to your sterility and microbial limits testing routine. Our range of validated solutions for the pharmaceutical microbiology laboratory support the production and quality assurance of sterile pharmaceuticals, vaccines, and biotechnology products.Complete Solutions, Complete Confidence:• Secure packaging: double bagged prior to autoclaving to preserve integrity of sterility media• Regulatory compliant: USP, JP, and EU harmonized media • Adaptable: a range of available fill volumes, media and bottle types to accommodate your sample sizes and performance testing needs• Quality guaranteed: Certificates of Analysis with every lot produced • Easy-to-use: simply ‘peel and pop’ bags for quick access to the media bottle• Multi-purpose media: suitable for general quality testing Customizable formats and media types are available. Please inquire.A highly nutritious versatile medium is recommended for general laboratory use. Due to the inclusion of both Tryptone and Soya Peptone, the medium will support a luxuriant growth of many fastidious organisms without the addition of serum.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Specifications

Cap Color Stopper, Crimp seal
Cap Type Stopper, Crimp seal
Content And Storage 2 to 25°C
Description Double Bagged Bottled Tryptone Soya Broth (TSB)
Format Double Bagged Serum Bottle
Product Type Microbiology Media
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.