Learn More
Invitrogen™ Donkey anti-Rat IgG (H+L) Secondary Antibody, FITC
Donkey Polyclonal Secondary Antibody
Brand:  Invitrogen™ A18740
Product Code. 15470724
  • 223.00€ / 2mg
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgG (H+L)
Polyclonal
FITC
Donkey
Affinity chromatography
RUO
Rat
Secondary Antibody
IgG
Flow Cytometry, Immunocytochemistry
1.5 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.8
Gamma Immunoglobins Heavy and Light chains.
2 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.