Learn More
Thermo Scientific™ Domed PCR Caps, strips of 12
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
57.75€ - 113.00€
Specifications
Color | Clear |
---|---|
Material | Polypropylene |
No. of Wells | 12 |
Format | Strips of 12 Domed Caps |
Product Type | PCR Plate Cap Strip |
Product Code | Brand | Color | Format | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Format | Quantity | Price | Quantity & Availability | |||
11584774
|
Thermo Scientific™
AB0852 |
Clear | Strips of 12 Domed Caps | 10 packs of 8 |
57.75€
Pack of 80 |
In Stock
Log in to see stock. |
|||
11594774
|
Thermo Scientific™
AB0853 |
Clear | Strips of 12 Domed Caps | 25 packs of 8 |
113.00€
Pack of 200 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.Highlights:
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Certified free from human DNA, RNases, and DNases.
Specifications
Clear | |
12 | |
PCR Plate Cap Strip | |
96-well Plate |
Polypropylene | |
Strips of 12 Domed Caps | |
Yes |