missing translation for 'onlineSavingsMsg'
Learn More

Serva Electrophoresis™ DNA Stain G

Brand:  Serva Electrophoresis™ 3980301

Encompass_Preferred

Product Code. 11417763

  • 84.70€ / 1mL
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Green
Fluorescent DNA Stain
Liquid
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SERVA DNA Stain G

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

Supplemental information
  • MIXTURE LIST-Contains : Hydrogen chloride

SDS
Documents

Documents

Certificates
Promotions

Promotions