missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMH-1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
DMH-1 is a selective inhibitor of bone morphogenic protein (BMP) type-I receptor activin receptor-like kinase 2 (ALK2) receptor (IC50 = 108 nM or 12.6 nM in in vitro kinase assays). DMH-1 exhibits 6- and 19-fold selectivity for ALK-2 over ALK-1 and ALK-3, respectively, and no significant inhibition of AMPK, ALK5, KDR (VEGFR-2) or PDGFRβ receptors. DMH-1 blocks BMP4-induced phosphorylation of Smads 1, 5 and 8 in HEK293 cells. Promotes neurogenesis in human induced pluripotent stem cells (iPSCs) when used in combination with SB 431542 (Cat. No. 1614). DMH-1 suppresses lung cancer cell proliferation, migration, invasion in vitro and reduces tumor growth in a mouse lung cancer xenograft model. DMH-1 inhibits cellular autophagy responses. DMH-1 induces intestinal differentiation in human intestinal organoids (hIOs) derived from human pluripotent stems cells (hPSCs).
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 380.44 |
Percent Purity | >98% |
Chemical Name or Material | DMH-1 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'