missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Serine, Non-Animal, ≥98.5%, Thermo Scientific Chemicals
156.00€ - 1244.00€
Chemical Identifiers
CAS | 302-84-1 |
---|---|
Molecular Formula | C3H7NO3 |
Molecular Weight (g/mol) | 105.093 |
MDL Number | MFCD00064223 |
InChI Key | MTCFGRXMJLQNBG-UHFFFAOYSA-N |
Synonym | dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid |
PubChem CID | 617 |
ChEBI | CHEBI:17822 |
IUPAC Name | 2-amino-3-hydroxypropanoic acid |
SMILES | C(C(C(=O)O)N)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15825188
|
Thermo Scientific
J21486.22 |
100 g | Plastic Bottle |
156.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15885328
|
Thermo Scientific
J21486.A1 |
1 kg | Plastic Bottle |
1244.00€
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
302-84-1 | |
105.093 | |
MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
617 | |
2-amino-3-hydroxypropanoic acid |
C3H7NO3 | |
MFCD00064223 | |
dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid | |
CHEBI:17822 | |
C(C(C(=O)O)N)O |
Specifications
∼240°C (decomposition) | |
C3H7NO3 | |
MFCD00064223 | |
dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid | |
C(C(C(=O)O)N)O | |
105.093 | |
CHEBI:17822 | |
≥98.5% | |
DL-Serine |
302-84-1 | |
CH2OHCHNH2COOH | |
1721402 | |
MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
2-amino-3-hydroxypropanoic acid | |
617 | |
105.09 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 206-130-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only