Learn More
DL-Proline, 99%, Thermo Scientific Chemicals
50.80€ - 480.00€
Chemical Identifiers
CAS | 609-36-9 |
---|---|
Molecular Formula | C5H9NO2 |
Molecular Weight (g/mol) | 115.132 |
MDL Number | MFCD00005250 |
InChI Key | ONIBWKKTOPOVIA-UHFFFAOYSA-N |
Synonym | dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275 |
PubChem CID | 614 |
ChEBI | CHEBI:26271 |
IUPAC Name | pyrrolidine-2-carboxylic acid |
SMILES | C1CC(NC1)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474504
|
Thermo Scientific Alfa Aesar
A13137.06 |
5 g |
50.80€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484504
|
Thermo Scientific Alfa Aesar
A13137.14 |
25 g |
163.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494504
|
Thermo Scientific Alfa Aesar
A13137.22 |
100 g |
480.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.
Solubility
Soluble in water and absolute alcohol. Insoluble in ether, butanol and isopropanol.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
609-36-9 | |
115.132 | |
ONIBWKKTOPOVIA-UHFFFAOYSA-N | |
614 | |
pyrrolidine-2-carboxylic acid |
C5H9NO2 | |
MFCD00005250 | |
dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275 | |
CHEBI:26271 | |
C1CC(NC1)C(=O)O |
Specifications
∼209°C (decomposition) | |
C5H9NO2 | |
80809 | |
dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275 | |
ONIBWKKTOPOVIA-UHFFFAOYSA-N | |
pyrrolidine-2-carboxylic acid | |
614 | |
115.13 | |
DL-Proline |
609-36-9 | |
MFCD00005250 | |
14,7780 | |
Soluble in water and absolute alcohol. Insoluble in ether,butanol and isopropanol. | |
C1CC(NC1)C(=O)O | |
115.132 | |
CHEBI:26271 | |
99% |
Safety and Handling
missing translation for 'einecsNumber' : 210-189-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only