missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Leucine, 99%
56.90€ - 641.00€
Chemical Identifiers
CAS | 328-39-2 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00063087 |
InChI Key | ROHFNLRQFUQHCH-UHFFFAOYNA-N |
Synonym | dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d |
PubChem CID | 857 |
ChEBI | CHEBI:25017 |
IUPAC Name | 2-amino-4-methylpentanoic acid |
SMILES | CC(C)CC(N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483254
|
Thermo Scientific Chemicals
A10590.18 |
50 g |
56.90€
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493254
|
Thermo Scientific Chemicals
A10590.30 |
250 g |
213.00€
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473254
|
Thermo Scientific Chemicals
A10590.0B |
1000 g |
641.00€
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
328-39-2 | |
131.18 | |
ROHFNLRQFUQHCH-UHFFFAOYNA-N | |
857 | |
2-amino-4-methylpentanoic acid |
C6H13NO2 | |
MFCD00063087 | |
dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d | |
CHEBI:25017 | |
CC(C)CC(N)C(O)=O |
Specifications
∼294°C (sublimation) | |
C6H13NO2 | |
636005 | |
dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d | |
CC(C)CC(N)C(O)=O | |
131.18 | |
CHEBI:25017 | |
99% | |
DL-Leucine |
328-39-2 | |
MFCD00063087 | |
14,5451 | |
ROHFNLRQFUQHCH-UHFFFAOYNA-N | |
2-amino-4-methylpentanoic acid | |
857 | |
131.18 | |
1.293 |
Safety and Handling
missing translation for 'einecsNumber' : 206-328-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only