Learn More
Thermo Scientific Chemicals DL-Histidine monohydrochloride monohydrate, 99%
30.90€ - 235.00€
Chemical Identifiers
CAS | 123333-71-1 |
---|---|
Molecular Formula | C6H12ClN3O3 |
Molecular Weight (g/mol) | 209.63 |
MDL Number | MFCD00151028 |
InChI Key | CMXXUDSWGMGYLZ-UHFFFAOYSA-N |
Synonym | dl-histidine; hypochlorous acid |
PubChem CID | 44828786 |
IUPAC Name | 2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride;hydrate |
SMILES | [H+].C1=C(NC=N1)CC(C(=O)O)N.O.[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11403227
|
Thermo Scientific Chemicals
A16165.06 |
5 g |
30.90€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413227
|
Thermo Scientific Chemicals
A16165.14 |
25 g |
83.70€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423227
|
Thermo Scientific Chemicals
A16165.22 |
100 g |
235.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
123333-71-1 | |
209.63 | |
CMXXUDSWGMGYLZ-UHFFFAOYSA-N | |
44828786 | |
[H+].C1=C(NC=N1)CC(C(=O)O)N.O.[Cl-] |
C6H12ClN3O3 | |
MFCD00151028 | |
dl-histidine; hypochlorous acid | |
2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride;hydrate |
Specifications
∼250°C (decomposition) | |
C6H12ClN3O3 | |
dl-histidine; hypochlorous acid | |
CMXXUDSWGMGYLZ-UHFFFAOYSA-N | |
2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride;hydrate | |
44828786 | |
99% |
123333-71-1 | |
MFCD00151028 | |
Insoluble in water. | |
[H+].C1=C(NC=N1)CC(C(=O)O)N.O.[Cl-] | |
209.63 | |
209.63 (191.62 Anhydrous) | |
DL-Histidine monohydrochloride monohydrate |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only