missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Histidine hydrochloride, monohydrate, 98%, Thermo Scientific Chemicals
68.40€ - 210.00€
Chemical Identifiers
CAS | 123333-71-1 |
---|---|
Molecular Formula | C6H10ClN3O2 |
Molecular Weight (g/mol) | 191.62 |
MDL Number | MFCD00064555 |
InChI Key | QZNNVYOVQUKYSC-JEDNCBNOSA-N |
Synonym | hydrogen dl-histidine chloride, 2-amino-3-1h-imidazol-5-yl propanoic acid; hydron; chloride, 2-azanyl-3-1h-imidazol-5-yl propanoic acid; hydron; chloride |
PubChem CID | 53395185 |
IUPAC Name | 2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride |
SMILES | [H+].[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10444121
|
Thermo Scientific Acros
411720250 |
25 g | Glass Bottle |
68.40€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10055770
|
Thermo Scientific Acros
411721000 |
100 g | Glass Bottle |
210.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123333-71-1 | |
191.62 | |
QZNNVYOVQUKYSC-JEDNCBNOSA-N | |
53395185 | |
[H+].[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O |
C6H10ClN3O2 | |
MFCD00064555 | |
hydrogen dl-histidine chloride, 2-amino-3-1h-imidazol-5-yl propanoic acid; hydron; chloride, 2-azanyl-3-1h-imidazol-5-yl propanoic acid; hydron; chloride | |
2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride |
Specifications
233.0°C | |
1ppm max. | |
10ppm max. | |
C6H10ClN3O2 | |
10ppm max. | |
1% max. | |
hydrogen dl-histidine chloride, 2-amino-3-1h-imidazol-5-yl propanoic acid; hydron; chloride, 2-azanyl-3-1h-imidazol-5-yl propanoic acid; hydron; chloride | |
[H+].[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O | |
191.62 | |
300ppm max. | |
96% min. (c=3, H2O, 430nm) 1cm cell | |
0.1% max. | |
DL-Histidine hydrochloride, monohydrate, 98% |
123333-71-1 | |
White | |
98% | |
Authentic | |
MFCD00064555 | |
(TLC) none | |
QZNNVYOVQUKYSC-JEDNCBNOSA-N | |
2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride | |
53395185 | |
209.64 | |
98% | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 229-266-8
missing translation for 'tsca' : TSCA
RUO – Research Use Only