missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-4-Chlorophenylalanine, 98%
63.10€ - 240.00€
Chemical Identifiers
CAS | 7424-00-2 |
---|---|
Molecular Formula | C9H10ClNO2 |
Molecular Weight (g/mol) | 199.63 |
MDL Number | MFCD00002601 |
InChI Key | NIGWMJHCCYYCSF-UHFFFAOYNA-N |
Synonym | fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro |
PubChem CID | 4652 |
IUPAC Name | 2-amino-3-(4-chlorophenyl)propanoic acid |
SMILES | NC(CC1=CC=C(Cl)C=C1)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10163060
|
Thermo Scientific Chemicals
157280050 |
5 g | Glass Bottle |
63.10€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10408130
|
Thermo Scientific Chemicals
157280250 |
25 g | Plastic Bottle |
240.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7424-00-2 | |
199.63 | |
NIGWMJHCCYYCSF-UHFFFAOYNA-N | |
4652 | |
NC(CC1=CC=C(Cl)C=C1)C(O)=O |
C9H10ClNO2 | |
MFCD00002601 | |
fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro | |
2-amino-3-(4-chlorophenyl)propanoic acid |
Specifications
>240°C | |
White | |
C9H10ClNO2 | |
ClC6H4CH2CH(NH2)CO2H | |
14, I, 608 | |
Solubility in water: slightly soluble. Other solubilities: slightly soluble in ethanol | |
NC(CC1=CC=C(Cl)C=C1)C(O)=O | |
199.63 | |
199.64 | |
0.5% max. (K.F.) | |
DL-4-Chlorophenylalanine, 98% |
7424-00-2 | |
97.5% min. (HPLC) | |
Authentic | |
MFCD00002601 | |
fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro | |
NIGWMJHCCYYCSF-UHFFFAOYNA-N | |
2-amino-3-(4-chlorophenyl)propanoic acid | |
4652 | |
98% | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 231-051-9
missing translation for 'rtecsNumber' : AY4390010
missing translation for 'tsca' : TSCA
RUO – Research Use Only