Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz Thiele, Straight, with Cooling Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05305114
 View more versions of this product
Product Code. 10468212
  • 846.00€ / Each
Explore more special offers
Description
Description
  • Receivers with straight adapter
  • With cooling jacket, graduated, with socket and cone NS 29/32
  • Vacuum connection: thread GL18
  • NS stopcock with solid glass or PTFE plug
  • Made of DURAN™ tubing
Specifications
Specifications
Duran Tubing, PTFE
100 mL
Socket and Cone NS 29/32