Learn More
Thermo Scientific™ Disposable IR Cards, 5x10cm, NaCl window, 9.5mm aperture
missing translation for 'orderingAttributeHoverText'
Quantity:
25 Pk.
5 Pk.
missing translation for 'unitSize'
Pack of 25
Pack of 5
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Size: 5 x 10cm
- NaCl window
- 9.5mm aperture
Spécification
Spécification
| Description | Disposable IR Cards |
| Quantity | 5 Pk. |