missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Disposable Gloves and Face Masks Zero Waste Box™
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Recycle Disposable Gloves and Face Masks using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
142.38€ - 316.18€
Specifications
Color | Beige, Green, Red, White |
---|---|
Disposable | Single-use |
Latex Free | Yes |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17805855
![]() |
Terracycle™
CS176-S-BE |
Small |
142.38€
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
17845935
![]() |
Terracycle™
CS176-M-BE |
Medium |
204.02€
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
17855935
![]() |
Terracycle™
CS176-L-BE |
Large |
316.18€
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
Description
Use this Zero Waste Box to recycle:
- Disposable protective face masks such as dust masks, FFP2 (or so-called ''N95''), FFP3 and surgical masks (as long as they don't come from a medical environment)
- Face shields and visors
- Disposable vinyl, nitrile and latex gloves
Approximate waste units accepted:
- Small: 520 units
- Medium: 1400 units
- Large: 2600 units
Do not send in:
- Medical waste
- This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, care homes, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Autoclaved material
- Gloves that are contaminated with materials considered hazardous or dangerous
- Fabric, homemade or paper masks or other types of protective garments
- Hairnets, earplugs, and lab coats or garments
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which presents a danger to the environment, or to people.
Specifications
Beige, Green, Red, White | |
Yes | |
Not Autoclavable | |
Recycling Box |
Single-use | |
Non-sterile | |
Cardboard, Plastic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title