missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neville and More™ Simplicity Lotion Pump Dispenser
HDPE Simplicity bottle with 24mm/410 White lotion pump.
Brand: Neville and More™ 024040250BEWTLPW
Additional Details : Weight : 0.23400kg
Description
- Dispense lotion, shampoo, conditioner, and many other viscous liquids.
- Constructed from HDPE.
Specifications
6/Pk. | |
HDPE | |
250 mL |
Pump Dispenser | |
White |