missing translation for 'onlineSavingsMsg'
Learn More

Disodium hydrogen citrate sesquihydrate, 99%, Thermo Scientific Chemicals

Product Code. 11457856
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Disodium hydrogen citrate sesquihydrate is a reagent employed in the investigation of mediatorless laccase cathodes as half-cell electrode under galvanostatic load by using a two-compartment set up. It is used in the extraction of organ/tissue sample.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disodium hydrogen citrate sesquihydrate is a reagent employed in the investigation of mediatorless laccase cathodes as half-cell electrode under galvanostatic load by using a two-compartment set up. It is used in the extraction of organ/tissue sample.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-05-4
Molecular Formula C12H18Na4O17
Molecular Weight (g/mol) 526.22
MDL Number MFCD00150445
InChI Key HGPVLOQNBSHYEI-UHFFFAOYNA-J
Synonym unii-cz1032cekr, cz1032cekr, sodium citrate dibasic sesquihydrate, disodium citrate sesquihydrate, disodium hydrogen citrate 1.5 h2o, sodium hydrogen citrate sesquihydrate, citric acid, disodium salt, sesquihydrate, tetrasodium, 3-carboxy-3-hydroxypentanedioate, trihydrate, sodium citrate dibasic sesquihydrate, purum p.a t, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, disodium salt, hydrate 2:3
PubChem CID 117063816
IUPAC Name tetrasodium;2-(carboxymethyl)-2-hydroxybutanedioate;trihydrate
SMILES C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 149°C (decomposition)
Quantity 5000 g
Solubility Information Soluble in water.
Formula Weight 263.11 (236.09 Anhydrous)
Percent Purity 99%
Chemical Name or Material Disodium hydrogen citrate sesquihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.