Learn More
Disodium hydrogen citrate sesquihydrate, 99%, Thermo Scientific Chemicals
Description
Disodium hydrogen citrate sesquihydrate is a reagent employed in the investigation of mediatorless laccase cathodes as half-cell electrode under galvanostatic load by using a two-compartment set up. It is used in the extraction of organ/tissue sample.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Disodium hydrogen citrate sesquihydrate is a reagent employed in the investigation of mediatorless laccase cathodes as half-cell electrode under galvanostatic load by using a two-compartment set up. It is used in the extraction of organ/tissue sample.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 149°C (decomposition) |
Quantity | 1000 g |
Solubility Information | Soluble in water. |
Formula Weight | 263.11 (236.09 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | Disodium hydrogen citrate sesquihydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.