Learn More
Brady™ Directional Arrow Tape
Product Code. 12787916
Click to view available options
Dimensions (LxW):
33 m x 100 mm
33 m x 25 mm
33 m x 50 mm
Color:
Black on Gray
Black on Ochre
Black on Orange
Black on White
Black on Yellow
White on Black
White on Blue
White on Green
White on Purple
White on Red
Unit Size:
1 roll
Product Code. 12787916
Brand: Brady™ 275127
Only null left
Add to basket
Add to basket
Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
  • Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
Specifications
Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Directional Arrow Tape
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Blue
Finish Gloss
Format Tape
Material Polyester
Product Type Pipe Marker
For Use With (Application) Pipe and valve marking
No. per Pack 1300 Markers/Roll
Quantity 1300 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (LxW) 33 m x 100 mm
Length (English) 108.27 ft.
Length (Metric) 33 m
Width (English) 3.94 in.
Width (Metric) 100 mm
Alternate Part Number ARROWTAPE:WHITE/BLUE100MMX33M