missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diprotin A, >98%, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Dipeptidyl peptidase IV (DPP-IV) inhibitor. Inhibits hydrolysis of incretins, regulating blood glucose levels. Induces Src and vascular endothelium-cadherin phosphorylation, increasing vascular permeability in the retina. Also increases number of viable phenotypic long term repopulating hematopoietic stem cells when harvesting mouse bone marrow in aerobic conditions.
Specifications
Specifications
CAS | 90614-48-5 |
Molecular Formula | C17H31N3O4 |
Linear Formula | C17H31N3O4 |
Quantity | 10 mg |
Synonym | diprotin a, ile-pro-ile, isoleucylprolylisoleucine, n-1-l-isoleucyl-l-prolyl-l-isoleucine, l-isoleucyl-l-prolyl-l-isoleucine, l-isoleucine, n-1-l-isoleucyl-l-prolyl, 2s,3s-2-2s-1-2s,3s-2-amino-3-methylpentanoyl pyrrolidin-2-yl formamido-3-methylpentanoic acid, h-ile-pro-ile-oh, spectrum2_001480, spectrum3_001838 |
Solubility Information | Soluble to 100mM in water |
InChI Key | JNTMAZFVYNDPLB-PEDHHIEDSA-N |
SMILES | CCC(C)C(C(=O)N1CCCC1C(=O)NC(C(C)CC)C(=O)O)N |
IUPAC Name | (2S,3S)-2-[[(2S)-1-[(2S,3S)-2-amino-3-methylpentanoyl]pyrrolidine-2-carbonyl]amino]-3-methylpentanoic acid |
Molecular Weight (g/mol) | 341.452 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'