Learn More
Esculape Laboratoire™ Diphoterine Eye Wash
Use Esculape Laboratoire™ Diphoterine Eye Wash to limit the extent of the burns and lesions by restricting the extent and severity of the lesions.
Brand:  Esculape Laboratoire™ 8205020
Product Code. 13418786
  • 78.00€ / 50mL
Edge
Explore more special offers
Description
Description
  • Sterile
  • Ergonomic eyecup facilitates eye opening.
  • Soft and autonomous rinsing without pressure
  • Sealed to assure the integrity of the system
  • Waterproof and compact EN 15454-4. EC certification
Specifications
Specifications
Eyewash Solution
Sterile
50 mL