Learn More
Thermo Scientific™ Dionex™ Fused Silica Tubing
Product Code. 11332123 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Inner:
10 μm
20 μm
25 μm
30 μm
50 μm
75 μm
100 μm
150 μm
200 μm
Length (Metric):
2.5 m
5 m
Unit Size:
Each
Product Code. 11332123
Brand: Thermo Scientific™ 160476
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.
Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LC-Packings Ultimate Capillary/Nano HPLC
Material Fused Silica
Type Capillary Tubing
Length (Metric) 5 m
Diameter (Metric) Inner 25 μm
Diameter (Metric) Outer 280 μm