Learn More
Dimethylglyoxime, disodium salt octahydrate, 99%, Thermo Scientific Chemicals
Dimethylglyoxime, disodium salt octahydrate, 99%, C4H6N2Na2O2.8H2O, CAS Number-75006-64-3, c4h6n2o2.2na.8h2o, dimethylglyoxime di-sodium salt octahydrate, dimethylglyoxime disodium salt octahydrate, sodium dimethylglyoxime octahydrate, 1kg, White to Beige, 262-523-2, 304.19
62.21€ - 432.15€
Chemical Identifiers
CAS | 75006-64-3 |
---|---|
Molecular Formula | C4H22N2Na2O10 |
Molecular Weight (g/mol) | 304.20 |
MDL Number | MFCD00150288 |
InChI Key | IJJDRGUPRATWQX-UHFFFAOYSA-L |
Synonym | dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate |
PubChem CID | 71310106 |
SMILES | O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10209700
|
Thermo Scientific Acros
408341000 |
100 g | Glass bottle |
62.21€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10146990
|
Thermo Scientific Acros
408342500 |
250 g | Glass bottle |
131.66€
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10136963
|
Thermo Scientific Acros
408340010 |
1 kg | Glass bottle |
432.15€
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75006-64-3 | |
304.20 | |
IJJDRGUPRATWQX-UHFFFAOYSA-L | |
71310106 |
C4H22N2Na2O10 | |
MFCD00150288 | |
dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate | |
O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-] |
Specifications
75006-64-3 | |
White to Beige | |
Authentic | |
C4H22N2Na2O10 | |
MFCD00150288 | |
(50mg/ml,50% ethanol) Clear solution | |
O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-] | |
304.20 | |
304.19 | |
Crystalline Powder |
>300.0°C | |
46% to 49% | |
99% | |
CH3C(NONa)C(NONa)CH3·8H2O | |
dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate | |
IJJDRGUPRATWQX-UHFFFAOYSA-L | |
disodium;2-N,3-N-dioxidobutane-2,3-diimine;octahydrate | |
71310106 | |
99% | |
Dimethylglyoxime, disodium salt octahydrate, 99% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 262-523-2
missing translation for 'tsca' : TSCA
- Dimethylglyoxime, disodium salt octahydrate
RUO – Research Use Only