Learn More
Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor
Accurately dispense media into suitable containers according to a set dilution using this gravimetric diluter which allows the use of two diluents.
6020.32€ - 8460.00€
Specifications
For Use With | Dry-Bags |
---|---|
Includes | Automated Dilutor Dual Pump unit only |
Material | Stainless Steel (Case) |
Overall Length (Metric) | 41.5 cm |
Overall Width (Metric) | 31 cm |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Prix | Quantité et disponibilité | |||||
16479713
|
Thermo Scientific™
DB4100A |
DiluFlux Pro-3 Automated Gravimetric Dilutor w/ simple (non-robotic) dispensing arm |
6020.32€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16489713
|
Thermo Scientific™
DB4150A |
DiluFlux Pro-5 Automated Gravimetric Dilutor w/ robotic dispensing arm |
8460.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Accurately dispense broth medium into suitable containers for food testing using the Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor. DiluFlux Pro Dilutors are compact, easy-to-use, and designed to deliver precise quantities of diluents rapidly with the touch of a button, so that you can work more confidently and accurately. When combined with the Thermo Scientific™ Peristaltic Pump and Thermo Scientific™ Homogenizer, your lab bench will be equipped with a flexible automated culture media solution to increase your productivity.
- Intuitive: simply press two keys, “tare” and “go”, and the DiluFlux Pro Dilutor will automatically dilute your sample to the desired dilution factor
- Accurate: featuring Watson-Marlow™ peristaltic pumps, high-tech flow regulators, and top of the line weighing platforms for trusted reproducibility and repeatability
- Versatile: compatible with many prepared media formats, the easy-to clean, stainless steel DiluFlux Pro Dilutor is an ideal fit for any food testing laboratory
- High-quality: the DiluFlux Pro Dilutors are ISO 7218, ISO 6887-1, FDA BAM-compliant and are made in France under ISO 9001:2008 certification
Spécification
Dry-Bags | |
Stainless Steel (Case) | |
31 cm | |
100/240 V |
Automated Dilutor Dual Pump unit only | |
41.5 cm | |
100/127 V 220/240 V 50/60 Hz | |
50/60 Hz |