Learn More
TFA Dostmann™ Digital Indoor Thermo-Hygrometer
An excellent choice for the monitoring of indoor temperature and humidity.
Brand:  TFA Dostmann™ 30.5015.02
Product Code. 11882881
  • 29.05€ / Each
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Description
  • Helps maintain a healthy indoor climate
  • Permanent indication of daily max. and min. values with auto reset
  • Additional max/min memory with manual reset
Specifications
Specifications
Thermohygrometer
-10°C to +70°C
Digital
134 mm
1.5 V
Thermo-hygrometer, battery, instruction manual
20 to 99%
1 x AAA 1.5 V
91 mm
100 g
For inside and outside measurement
CE, EK-EL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products