missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Sabouraud Dextrose Broth

Product Code. 11788832
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11788832

Brand: BD 238230

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Used in qualitative procedures for culturing yeasts, molds and aciduric microorganisms. BD™ Difco™ Sabouraud Dextrose Broth meets United States Pharmacopeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP) performance specifications.

Sabouraud Dextrose Broth is used for culturing yeasts and molds in cosmetics. The USP recommends the use of Sabouraud Dextrose Broth when isolating Candida albicans from nonsterile pharmaceutical products.

  • Sabouraud dextrose media are peptone media supplemented with dextrose to support the growth of fungi
  • Peptones are sources of nitrogenous growth factors
  • The carbohydrate provides an energy source for the growth of microorganisms
  • pH (3.0% solution, 25°C): 5.6 ± 0.2
  • Displays cultural response to: Aspergillus brasiliensis (niger) 16404, Candida albicans 10231, Lactobacillus casei 9595, Saccharomyces cerevisiae 9763, Candida albicans 10231
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, EP, JP, USP
Color Light beige
Content And Storage 2 to 25°C
Form Dehydrated Powder
Media Type Sabouraud Dextrose Broth
Product Type Microbiology Media
Formulation Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 5g/L, Dextrose 20g/L
For Use With (Application) Cultivation of Yeasts, molds and aciduric microorganisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.