missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylether a.r. Stabilized with 5 to 7 ppm BHT For Laboratory Use, Chem Lab
Click to view available options
Diethylether a.r. Stabilized with 5 to 7 ppm BHT For Laboratory Use, 1 L
Specifications
Specifications
Melting Point | -116°C |
CAS | 60-29-7 |
Color | Colorless |
Acidity | < 0.0002 meq/g |
Density | 0.71 g/mL |
Boiling Point | 34°C |
Quantity | 1 L |
Flash Point | -40 °C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Show More |
Safety and Handling
Product Identifier
- Diethylether
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 1
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P403+P235-Store in a well-ventilated place. Keep cool.
- MIXTURE LIST-Contains : Diethylether
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction