missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, ACS Reagent, Stabilized, Honeywell Burdick & Jackson™
Click to view available options
195 L
Specifications
Specifications
Melting Point | -116°C (1,013 hPa) |
CAS | 60-29-7 |
Color | Colorless |
Molecular Formula | C4H10O |
Linear Formula | (CH3CH2)2O |
Beilstein | 1696894 |
Synonym | Ether; Ethyl ether |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
SMILES | CCOCC |
IUPAC Name | ethoxyethane |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction