missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riedel-de Haën Dichloromethane with Amylene
Click to view available options
Quantity:
7 L
18 L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals.
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
pH | Not applicable |
Melting Point | -97°C |
CAS Min % | ≥99.00% |
Color | Colorless |
Density | 1.32 to 1.33 g/cm3 (20°C) |
Boiling Point | 39°C to 41°C (1.013 hPa) |
Quantity | 18 L |
Flash Point | Not applicable |
Packaging | Returnable Stainless Steel Can |
Physical Form | Liquid |
Show More |
Safety and Handling
Product Identifier
- Dichloromethane
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'