missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl 5-aminoisophthalate, 96%, Thermo Scientific™

Product Code. 11333626
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152699-63-3
Molecular Formula C22H19NO4
Molecular Weight (g/mol) 361.397
MDL Number MFCD03093065
InChI Key VDJWRMQQXQBYIO-UHFFFAOYSA-N
Synonym dibenzyl 5-aminoisophthalate, 1,3-dibenzyl 5-aminobenzene-1,3-dicarboxylate, acmc-20alb1, 3,5-dibenzyloxycarbonylaniline, 3,5-dibenzyloxycarbonyl-aniline, 5-amino-isophthalic acid dibenzyl ester, phenylmethyl 5-amino-3-benzyloxycarbonyl benzoate, 1,3-benzenedicarboxylicacid, 5-amino-, 1,3-bis phenylmethyl ester
PubChem CID 7023592
IUPAC Name dibenzyl 5-aminobenzene-1,3-dicarboxylate
SMILES C1=CC=C(C=C1)COC(=O)C2=CC(=CC(=C2)N)C(=O)OCC3=CC=CC=C3

Specifications

Melting Point 116°C to 118°C
Quantity 5 g
Beilstein 744276
Formula Weight 361.4
Percent Purity 96%
Chemical Name or Material Dibenzyl 5-aminoisophthalate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'