missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ DexPure Nitrile Gloves, Powder-Free
Click to view available options
Size:
10
9
Large
Small
Description
- Nitrile construction for chemical and mechanical resistance.
- Patterned finish for good grip.
- Certified for food contact and treated to prevent bacteria.
- Suitable for the following:
- Agrifood industry.
- Electronics and complex assembly.
- Graphic industry.
- Painting in the automotive industry.
- Pharmaceutical laboratories.
- Chemical handling.
- Lightly powdered to facilitate donning and doffing.
- Single use.
- Alternative for latex allergy sufferers.
Specifications
Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 374-2003 Chemical hazards, EN 455-1&2 Medical Standards, European directive N°1934/2004:Food contact, C Category PPE 2, EC Certificate Number 0075/797/162/12/08/0490, EC Attestation Number 0075/797/162/12/08/0490 |
Classification | Dexterity: 5 |
Color | Blue |
Disposable | Single-use |
Exterior Finish | Patterned |
Material | Nitrile |
Powdered | Powdered |
Product Type | Gloves |
Size | Large |
Thickness (Metric) | 0.1 mm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'