Learn More
KGW Isotherm™ Dewar Insulated Carrying Flasks
For the storage and transport of temperature-sensitive goods such as dry ice. KGW Isotherm™ Dewar Insulated Carrying Flasks feature a robust design made from borosilicate glass 3.3.
Brand:  KGW Isotherm™ 1212
 View more versions of this product
Product Code. 10312861
  • 380.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Cooling of small samples
  • Silver-plated and evacuated
  • Flexible applications in the laboratory
  • Includes handle made from aluminum
  • Includes insulated lid, held by two spring clips
  • Available with either blue-coated metal cover or stainless steel cover
Specifications
Specifications
Blue-coated Metal Cover
Cooling of small samples
245mm
2 L
Borosilicate Glass
160mm
170mm
Insulated lid, held by two spring clips, handle
138 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
KGW Isotherm™ Dewar Insulated Carrying Flasks > Capacity: 2000mL; Cover: Blue coated; Inner Height: 170mm; Inner Diameter: 138mm