missing translation for 'onlineSavingsMsg'
Learn More

Smeg™ Deterliquid I2

Liquid alkaline detergent comes in convenient 5 L can.

Brand:  Smeg™ DETERLQI2


Product Code. 16440815

  • 110.00€ / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Liquid alkaline detergent comes in convenient 5 L can.

  • Contains sodium hydroxide, phosphonic-carboxylic acid, sodium hypochlorite
Specifications

Specifications

Can
1.32 gal.
5 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • deterliquid I2

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH206-Warning

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.

Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,sodium hypochlorite, tetrasodium-phosphonoethane-1,2-dicarboxylate, hexasodium-

SDS
Documents

Documents

Certificates
Promotions

Promotions