missing translation for 'onlineSavingsMsg'
Learn More

Smeg™ Deterliquid C2

Liquid alkaline detergent washes steel at up to 95°C.

Brand:  Smeg™ DETERLQC2


Product Code. 16256631

  • 152.00€ / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Strong universal liquid alkaline detergent comes in convenient 5 L can.

  • Phosphate-free
  • For washing steel up to 95°C
  • Also suitable for washing rubber and plastic at 60°C
  • Contains sodium silicate, polyacrylate, potassium hydroxide
Specifications

Specifications

1.32 gal.
5 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P351-Rinse cautiously with water for several minutes.

Supplemental information
  • MIXTURE LIST-Contains : soudium silicate, polyacrylate, potassium hydroxide

SDS
Documents

Documents

Certificates
Promotions

Promotions