missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alla France™ Density Hydrometers with Thermometer
Alla France™ Density Hydrometer with Thermometer is designed to measure the relative density of a liquid.
Brand: Alla France™ 3011TP100/20-qp
This item is not returnable.
View return policy
Description
- Calibration temperature 20°C
- 1.000-1.100 - General Purpose Thermo-hydrometer 100 g/mL
- For use in the distillery and wine industry
Specifications
Hydrometer | |
0.001 | |
Thermometer |
±0.001 | |
320 mm | |
Range: 1.0 to 1.1 g/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction