Learn More
OHAUS™ Defender™ 3000 Washdown - I-D33 Bench Scale (EU), Certified
Stainless Steel Bench Scales for Basic Industrial Washdown Applications
Brand:  OHAUS™ 30685136
 View more versions of this product
Product Code. 17515825
  • 1015.00€ / Each
Explore more special offers
Description
Description
  • Results are clearly shown using the large, bright backlit LCD display.
  • In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
  • This industrial scale features a 304 stainless steel platform and frame, IP67 load cell.
  • Indicator features an IP66 washdown protected 304-stainless steel housing with mounting bracket.
  • Features software for Parts Counting, Checkweighing and Totalization.
  • Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 Hours with Rechargeable Battery
Standard RS232 and optional USB Device or Ethernet
60 kg
355 x 305 mm
D33XW60C1R6EU
-10°C to +40°C, 85%RH, non-condensing
491 x 305 x 569 mm
AC Adapter (Included);Rechargeable Battery (Included)
304-grade stainless steel platform and frame, stainless steel IP66 indicator, IP67 load cell, non-slip adjustable rubber feet
7 Segment LCD with Multicolor Backlight
20 g
IP66
Weighing, Checkweighing, Counting, Totalization.
12.4 kg