Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale
Shop All Ohaus Products
Click to view available options
Capacity:
15 kg
30 kg
60 kg
150 kg
300 kg
600 kg
Model:
D33P150B1L2
D33P150B1L5
D33P150B1X2
D33P150B1X5
D33P15B1R1
D33P15B1R5
D33P300B1V3
D33P300B1X2
D33P300B1X5
D33P30B1R1
D33P30B1R5
D33P600B1V3
D33P60B1L2
D33P60B1L5
D33P60B1R1
D33P60B1R5
Readability:
2 g
5 g
10 g
20 g
50 g
100 g
Dimensions (L x W x H):
489 x 305 x 542 mm
646 x 305 x 130 mm
686 x 420 x 896 mm
785 x 500 x 896 mm
842 x 420 x 134 mm
941 x 500 x 150 mm
933 x 600 x 1195 mm
Unit Size:
Each
Beschreibung
- Results are clearly shown using the large, bright backlit LCD display.
- In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
- This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
- Indicator features a stainless steel housing and mounting bracket.
- Features software for Parts Counting, Checkweighing and Totalization.
- Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Spezifikation
Spezifikation
| Battery Life | 100 Hours with Rechargeable Battery;360 Hours with Disposable Batteries |
| Battery Type | 6 C (LR14) Batteries (Not Included);AC Adapter (Included);Rechargeable Battery (Not Included) |
| Connection Type | Standard RS232 and optional USB Device or Ethernet |
| Construction | ABS plastic indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet |
| Capacity | 300 kg |
| Display Type | 7-segment Multi-color Backlit LCD |
| Dimensions (LxW) Platform | 650 x 500 mm |
| Readability | 50 g |
| Model | D33P300B1X5 |
| Temperature (Metric) Operating | -10°C to +40°C, 85%RH, non-condensing |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur