missing translation for 'onlineSavingsMsg'
Learn More

DEET-(diethyl-d10), Honeywell Fluka™

Product Code. 15951051
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

CAS 1215576-01-4
Packaging Glass Bottle
Quantity 10 mg
Molecular Formula C12D10H7NO
MDL Number MFCD28118921
Molecular Weight (g/mol) 201.33
Grade Analytical Standard
Density 1 g/cm3
Boiling Point 110°C
For Use With (Application) Acaricides and Insecticides
Physical Form Liquid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'