Learn More
Fisherbrand™ Decapping Heads for Programmable Electronic High Power Crimp Station
Description
- Decapping Head for Alumnium/Steel Caps
- Not adjustable
Specifications
20mm Decapping Head | |
Plastic, Steel | |
20 mm Alumnium Caps |
20 mm | |
Not Autoclavable |