missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBPA Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-71827CL1
This item is not returnable.
View return policy
Description
DBPA Polyclonal antibody specifically detects DBPA in Human samples. It is validated for Immunoprecipitation,Western BlotSpecifications
DBPA | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
8531 | |
Store at 4°C in the dark. Do not freeze. | |
IgG |
Immunoprecipitation, Western Blot | |
CoraFluor 1 | |
cold shock domain protein A, Cold shock domain-containing protein A, cold-shock domain protein A, CSDA1, dbpA, DBPAcold-shock domain containing A1, DNA-binding protein A, Single-strand DNA-binding protein NF-GMB, ZO-1-associated nucleic acid-binding protein, ZONAB | |
The immunogen for this product maps to a region between residue 250 and 300 of human ZO-1-associated nucleic acid-binding protein using the numbering given in entry NP_003642.3 (GeneID 8531). | |
0.1 mL | |
Primary | |
Human | |
Purified |