missing translation for 'onlineSavingsMsg'
Learn More

DBPA Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-71827AF350

Product Code. 18485873

  • 639.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

DBPA Polyclonal antibody specifically detects DBPA in Human samples. It is validated for Western Blot
Specifications

Specifications

DBPA
Polyclonal
Western Blot
cold shock domain protein A, Cold shock domain-containing protein A, cold-shock domain protein A, CSDA1, dbpA, DBPAcold-shock domain containing A1, DNA-binding protein A, Single-strand DNA-binding protein NF-GMB, ZO-1-associated nucleic acid-binding protein, ZONAB
The immunogen for this product maps to a region between residue 250 and 300 of human ZO-1-associated nucleic acid-binding protein using the numbering given in entry NP_003642.3 (GeneID 8531).
0.1 mL
Primary
Human
Purified
Western Blot
Alexa Fluor 350
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
8531
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions