Learn More
3M™ Secure Click™ Replaceable Particulate Filter
3M™ Secure Click™ Replaceable Particulate Filter is CE approved for environments containing certain solid and liquid particles.
98.80€ - 126.00€
Specifications
Certifications/Compliance | CE Approved |
---|---|
Connection Type | Secure Click™ |
Product Type | Replacement Filter |
Product Code | Brand | Filter Type | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | Model | Price | Quantity & Availability | ||||
17207216
|
3M™
7100197286 |
P3R | D7925 |
107.00€
20 pairs |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
17297206
|
3M™
7100197285 |
P2R | D7935 |
126.00€
20 pairs |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
16446390
|
3M™
7100197286 |
P3R | D7925 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
16436390
|
3M™
7100197285 |
P2R | D7935 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
Use our filters with 3M™ Secure Click™ Respirators 800 Series, 3M™ Secure Click™ Cartridges D8000 Series and 3M™ Filter Retainer D701.Specifications
CE Approved | |
Secure Click™ | |
Replacement Filter |