missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Leucine, 99%
38.20€ - 81.20€
Chemical Identifiers
CAS | 328-38-1 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00063088 |
InChI Key | ROHFNLRQFUQHCH-RXMQYKEDSA-N |
Synonym | d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin |
PubChem CID | 439524 |
ChEBI | CHEBI:28225 |
IUPAC Name | (2R)-2-amino-4-methylpentanoic acid |
SMILES | CC(C)C[C@@H](N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10635852
|
Thermo Scientific Chemicals
226900025 |
2.5 g | Glass Bottle |
38.20€
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10357980
|
Thermo Scientific Chemicals
226900100 |
10 g | Glass Bottle |
81.20€
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
328-38-1 | |
131.18 | |
ROHFNLRQFUQHCH-RXMQYKEDSA-N | |
439524 | |
(2R)-2-amino-4-methylpentanoic acid |
C6H13NO2 | |
MFCD00063088 | |
d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin | |
CHEBI:28225 | |
CC(C)C[C@@H](N)C(O)=O |
Specifications
328-38-1 | |
White | |
99% | |
Authentic | |
10ppm max. | |
04, 446 | |
d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin | |
CC(C)C[C@@H](N)C(O)=O | |
0.1% max. | |
131.18 | |
CHEBI:28225 | |
≥98.5% | |
Crystalline Powder, Crystals, or Flakes |
2ppm max. | |
10ppm max. | |
C6H13NO2 | |
(CH3)2CHCH2CH(NH2)CO2H | |
MFCD00063088 | |
0.2% max. | |
ROHFNLRQFUQHCH-RXMQYKEDSA-N | |
(2R)-2-amino-4-methylpentanoic acid | |
-14.9° to -16° (20°C, 589nm) (c=4, 6N HCl) | |
439524 | |
131.17 | |
−15.45° (20°C c=4,6N HCl) | |
D-Leucine, 99% |
Safety and Handling
missing translation for 'einecsNumber' : 206-327-7
missing translation for 'rtecsNumber' : OH2840000
missing translation for 'tsca' : TSCA
RUO – Research Use Only