Learn More
Thermo Scientific Chemicals D-Glutamic acid, 99+%
49.10€ - 570.00€
Chemical Identifiers
CAS | 6893-26-1 |
---|---|
Molecular Formula | C5H9NO4 |
Molecular Weight (g/mol) | 147.13 |
MDL Number | MFCD00063112 |
InChI Key | WHUUTDBJXJRKMK-GSVOUGTGSA-N |
Synonym | d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid |
PubChem CID | 23327 |
ChEBI | CHEBI:15966 |
IUPAC Name | (2R)-2-aminopentanedioic acid |
SMILES | C(CC(=O)O)C(C(=O)O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482257
|
Thermo Scientific Chemicals
A14191.06 |
5 g |
49.10€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492257
|
Thermo Scientific Chemicals
A14191.14 |
25 g |
171.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402267
|
Thermo Scientific Chemicals
A14191.22 |
100 g |
570.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Glutamic acid acts as a superior to alpha ketoglutarate. It helps in the growth of Lactobacillus arabinosus. It can inhibit the IgE binding to peanut allergens. It is widely used in pharmaceuticals and foods.
Solubility
Soluble in water (8.88 mg/mL) at 25°C.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Chemical Identifiers
6893-26-1 | |
147.13 | |
WHUUTDBJXJRKMK-GSVOUGTGSA-N | |
23327 | |
(2R)-2-aminopentanedioic acid |
C5H9NO4 | |
MFCD00063112 | |
d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid | |
CHEBI:15966 | |
C(CC(=O)O)C(C(=O)O)N |
Specifications
199°C (decomposition) | |
C5H9NO4 | |
1723800 | |
d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid | |
WHUUTDBJXJRKMK-GSVOUGTGSA-N | |
(2R)-2-aminopentanedioic acid | |
23327 | |
147.13 | |
−30° (c=10 in 2N HCl) |
6893-26-1 | |
MFCD00063112 | |
14,4469 | |
Soluble in water (8.88mg/mL) at 25°C. | |
C(CC(=O)O)C(C(=O)O)N | |
147.13 | |
CHEBI:15966 | |
≥99% | |
D-Glutamic acid |
Safety and Handling
missing translation for 'einecsNumber' : 230-000-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only